| Related Categories | Eye Protection Equipment, Goggles, Labware, Safety Equipment |
| availability | not available in USA and Canada |
General use goggles with a clear polycarbonate lens that provides grade 1 impact protection as well as meets the British Standards for chemical splash, dusts and molten metals. BS2092.1CDM