| Related Categories | Bel-Art, Bel-Art Labels & Markers, General Labels, Label Tape, Labels and Markers, |
| L × W × H | 5 in. × 8 1/2 in. × 1/2 in. |
For use where additional weight is required to resist dispenser slippage when dispensing tape. Weight is epoxy coated steel with four rubber feet. Two threaded holes accept the screws provided to mount to tape dispenser Z115479.
Scienceware is a registered trademark of Bel-Art Products