ISBN-10 0198557914
ISBN-13 978-019855791-3
| Related Categories | Books and Software, Labware, Organic Chemistry Books More... |
| publication info | C. Willis, Oxford University Press, 1996, 96 pp., soft cover |
This practical book uses a wide range of examples in a versatile approach to the design of effective syntheses. Retrosynthetic analysis--a method of identifying simple starting materials for a synthesis--is introduced with emphasis on the importance of bond polarity and functional group interconversions. The next section discusses how an effective route to a target molecule containing more than one functional group can be followed, while later chapters review methods for the control of chemo-, regio-, and stereoselectivity, and include a discussion of protecting groups. Finally, four syntheses of pyrrolidine alkaloids are compared using previously described principles.
1. Introduction to synthesis
2. Retrosynthetic analysis I: The basic concepts
3. Retrosynthetic analysis II: Latent polarity and FGIs
4. Retrosynthetic analysis III: Strategy and planning in synthesis
5. Selectivity I: Chemoselectivity and protecting groups
6. Chemoselectivity II: Regioselectivity
7. Chemoselectivity III: Stereoselectivity
8. Selected organic synthesis
Glossary
Appendix: Synthon table