| Related Categories | Eye Protection Equipment, Eyeshields, Labware, Safety Equipment |
| availability | not available in EU |
| color | yellow temple arm |
| clear Lens |
• Rachet mechanism allows 12 different positions
• 5-position temple arm length adjustment
• One-piece, scratch resistant, clear PC lens
• Integral lens sideshields
• Choice of temple arm colors
• Meets ANSI Z87.1-1989 requirements