| Related Categories | Eye Protection Equipment, Faceshields, Labware, MSA, Manufacturer Browser, |
| material | clear acetate visor |
| mfr. no. | MSA, 488133 |
| availability | not available in EU |
| size | 8 in. × 16 in. × 0.04 in. |
• Lightweight, flexible headbands adjust to accommodate head sizes 6
• Effective against most flying fragments, particles, splashes, and similar hazards
• Optional nitrometer chin guard protects against upsweep of particles and splash
• Use with approved spectacles or goggles
• Assemblies comply with ANSI Z87.1
Order Defender + headgear assembly, visor and chin guard separately.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?