ISBN-10 0471963712
ISBN-13 978-047196371-4
| Related Categories | Books and Software, Labware, Organic Chemistry Books More... |
| publication info | H. Ulrich, John Wiley & Sons, Ltd, 1997, 514 pp., hard cover |
Chemistry and Technology of Isocyanates is a comprehensive book on isocyanate chemistry and technology. It highlights the industrial applications of diisocyanates in the manufacture of flexible and rigid foams, elastomers, coatings and adhesives; discusses ionomers used in water-based coatings, polymer networks and biomedical polymers; and reviews current and future environmental issues, including toxicity and safe handling of isocyanates, recycling of isocyanate derived polymers and monomers derived from natural products.
Monoisocyanates.
Diisocyanates.
Environmental Considerations.
Index.