| Related Categories | Eye Protection Equipment, Eyeshields, Labware, Safety Equipment |
| availability | not available in EU |
| color | amber Lens |
Polycarbonate lenses supported on black Resilium nylon frame, with a Soft-Fit nose bridge to prevent slippage. 100% UV protection. Meets ANSI Z87.1 standards.