Z670634 Aldrich

Whatman® Protran® nitrocellulose membranes

Circles - BA85, pore size 0.45 μm, diam. 25 mm, pack of 100 ea

DOWNLOAD MSDS (PDF)

Synonym: blotting membranes, nitrocellulose membrane, transfer membranes

Properties

Related Categories Blotting Membranes and Paper, Electrophoresis Supplies, Labware, Manufacturer Browser, Protran BA85 (0.45 μm pore size, binding capacity 80 μg/cm2),
mfr. no.   Whatman 10402506
  Whatman Article No., 28415097 (US reference)
packaging   pack of 100 ea
diam.   25 mm
pore size   0.45 μm pore size

Description

General description

Manufactured using 100% pure nitrocellulose to ensure the highest binding capacity. The surface properties of the membrane ensure superior signal-to-noise ratios without the need for stringent washing conditions. The proprietary Protran nitrocellulose formula is proven to provide excellent shelf life of proteins. Evidence shows proteins maintain molecular recognition activity for five years on Protran.

• Binding capacity, 75 - 110 μg/cm2
• High binding, low background
• Triton free matrix
• Universal membrane for blotting - isotopic, chemluminescent, colorimetric and fluorescent detection methods

Protran nitrocellulose (NC) membranes are the most frequently specified transfer media in the world for a wide range of applications. Protran membranes are manufactured using 100% pure nitrocellulose to ensure the highest binding capacity possible. Other membranes referred to as ‘nitrocellulose’ may actually contain large amounts of cellulose acetate which will lower the protein binding capacity. Protran membranes have the best handling strength of all pure nitrocellulose membranes.

They are compatible with a variety of detection methods, including isotopic, chemiluminescent (luminol-based), colorimetric and fluorescent. Unlike PVDF membranes, Protran nitrocellulose does not require a methanol pre-wetting step. This makes Protran the membrane of choice for proteins which prefer aqueous environments. Prior to transfer, the membrane is simply wetted in water, and then placed in the transfer buffer. No other pre-treatment steps are necessary.

High Binding, Low Background:
In addition to high binding capacity, Protran nitrocellulose membranes inherently have very low background. The superior surface properties of the membrane guarantee superior signal-to-noise ratios, without the need for stringent washing conditions.

High Retention of Small Proteins:

The 0.2 μm pore size of the Protran (BA83) nitrocellulose membrane has high surface area, ensuring binding of small proteins below 20 kD by reducing ‘blow-through’. The 0.45 μm pore size membrane (BA85) is ideal for larger molecular weight samples. BA79, with 0.1 μm pore size, is the membrane of choice for smaller proteins below 7 kD. A unique benefit of the proprietary Protran nitrocellulose formula is the proven shelf life of bound proteins. Empirical evidence shows that proteins maintain molecular recognition activity for five years on Protran.

Protran Blotting Sandwiches:

A pre-cut nitrocellulose membrane and 2 sheets of 3MM Chr blotting paper are prepackaged into a sandwich to help save time. This is made with our Protran BA83 or BA85, the highest quality NC membranes available for blotting.

Other Notes

Field of Use : Any microbiological or biotechnological application, including, but not limited to application of membranes for diagnostic, medical, biological, pharmaceutical, consumer product and food and beverage purposes.

Legal Information

Protran is a registered trademark of GE Healthcare

Whatman is a registered trademark of GE Healthcare

Price and Availability

Customers Also Viewed

Nitrocellulose membranes for blotting

pore size 0.45 μm, size 15 cm × 15 cm

Whatman<SUP>®</SUP> Protran<SUP>®</SUP> nitrocellulose membranes

Circles - BA83, pore size 0.2 μm, diam. 82 mm

Whatman<SUP>®</SUP> Protran<SUP>®</SUP> nitrocellulose membranes

Circles - BA85, pore size 0.45 μm, diam. 25 mm, pack of 1000 ea

Whatman<SUP>®</SUP> Protran<SUP>®</SUP> nitrocellulose membranes

Circles - BA85, pore size 0.45 μm, diam. 82 mm

Whatman<SUP>®</SUP> Protran<SUP>®</SUP> nitrocellulose membranes

Rolls - BA85, pore size 0.45 μm, W × L 30 cm × 3 m

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?