Z671355 Aldrich

Whatman® UniPrep® syringeless filters

PVDF membrane, pore size 0.45 μm

DOWNLOAD MSDS (PDF)

Synonym: Whatman filter, Whatman filters, syringe filters

Properties

Related Categories Filtration Supplies, HPLC Sample Preparation, Labware, Manufacturer Browser, Syringeless Filters,
material   PVDF membrane
  polypropylene housing
sterility   non-sterile
feature   suggested use low protein binding
mfr. no.   Whatman Article No., 28421650 (US reference)
  Whatman UN113UAQU
capacity   1-5 mL
hold-up volume   50 mL
pore size   0.45 μm

Description

General description

• Integral storage vial saves time and minimises laboratory waste
• Built-in prefilters means even difficult samples are quick and easy to prepare

UniPrep syringless filters are pre-assembled filtration devices for the filtration and storage od laboratory samples. These devices are quick and easy-to-use and feature a plunger, filter and vial in one unit.

The design incorporates a prefilter and a membrane into the tip of the plunger. When the filter plunger is pressed through the liquid placed in the test tube, positive pressure forces the filtrate up into the reservoir of the filter-lunger.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Uniprep is a registered trademark of Whatman International Ltd.

Whatman is a registered trademark of GE Healthcare

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?