| Related Categories | Cell Culture Supplies, Culture Dishes, Labware More... |
| packaging | pkg of 500 ea |
| diam. × deep | 50 mm × 20.4 mm |
Sterile high quality, disposable petri dishes, aseptically manufactured in clear polystyrene to the highest standards. Suitable for all applications including bacteriological.
Customers Also Viewed

diam. × H 100 mm × 25 mm, surface area size 57 cm2, Deep Petri with stacking ring; non-vented, Lab-Tek® petri dish
shallow 122, diam. 50 mm
TV 101VR20, diam. 90 mm
square, L × W 120 mm × 120 mm
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?