• USA Home
  • A15002 - 4-Acetyl-3,5-dimethyl-2-pyrrolecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
A15002 Aldrich

4-Acetyl-3,5-dimethyl-2-pyrrolecarboxylic acid

95%

  • CAS Number 2386-28-9

  • Empirical Formula (Hill Notation) C9H11NO3

  • Molecular Weight 181.19

  •  MDL number MFCD00022374

  •  PubChem Substance ID 24890548

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyrroles
InChI Key   SQFWNYAZHOVSPA-UHFFFAOYSA-N
assay   95%
mp   205-207 °C(lit.)

Description

Packaging

10, 50 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?