• USA Home
  • A33205 - Allyl isothiocyanate

EMAIL THIS PAGE TO A FRIEND
A33205 Aldrich

Allyl isothiocyanate

97%

Synonym: AITC, Oil of mustard

  • CAS Number 57-06-7

  • Linear Formula CH2=CHCH2NCS

  • Molecular Weight 99.15

  •  Beilstein Registry Number 773748

  •  MDL number MFCD00004822

  •  PubChem Substance ID 329770718

Purchase

Properties

InChI Key   ZOJBYZNEUISWFT-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.529(lit.)
bp   150 °C(lit.)
mp   −80 °C(lit.)
density   1.013 g/mL at 25 °C(lit.)

Description

Biochem/physiol Actions

Naturally occurring constituent of cruciferous vegetables that has anti-tumor and anti-oxidant properties. Induces the activity of phase II detoxification enzymes in the urinary bladder and, thus, may be chemoprotective against bladder cancer.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?