• USA Home
  • A72200 - 4-Aminophenyl disulfide

EMAIL THIS PAGE TO A FRIEND
A72200 Aldrich

4-Aminophenyl disulfide

95%

Synonym: 4,4′-Dithiodianiline

Purchase

Properties

InChI Key   MERLDGDYUMSLAY-UHFFFAOYSA-N
assay   95%
mp   77-78 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?