EMAIL THIS PAGE TO A FRIEND
A72502 Aldrich

4,4′-Oxydianiline

97%

Synonym: 4,4′-Diaminodiphenyl ether, 4-Aminophenyl ether

Purchase

Properties

vapor pressure   10 mmHg ( 240 °C)
InChI Key   HLBLWEWZXPIGSM-UHFFFAOYSA-N
assay   97%
mp   188-192 °C(lit.)

Description

Legal Information

DuPont product

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
BY7900000
Flash Point(F) 
426.2 °F
Flash Point(C) 
219 °C
Protocols & Articles

Articles

[JIM BALL TEST DOC] MIDA-protected Boronate Esters

The Suzuki-Miyaura cross-coupling reaction is one of the most important and highly utilized reactions in organic chemistry, with applications in polymer science as well as in the fine chemicals and p...
Josephine Nakhla
ChemFiles Volume 9 Article 1
Keywords: Aminations, Applications, Bromoborations, Building blocks, Catalysis, Chemfiles, Chromatography, Coupling reactions, Cross couplings, Cross metathesis, Cyclopropanations, Epoxidations, Industries, Ligands, Metathesis, Methods, Natural product synthesis, Olefin metathesis, Pharmaceutical, Polymer science, Protocols, Purification, Reductions, Reductive aminations, Stille coupling, Suzuki coupling, Suzuki reactions, Takai olefination, Transmetalation, Type, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?