EMAIL THIS PAGE TO A FRIEND
B103284 Aldrich

Butyraldehyde

99%

Synonym: Butanal

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C1 to C6, Carbonyl Compounds, Chemical Synthesis,
vapor density   2.5 (vs air)
vapor pressure   90 mmHg ( 20 °C)
  91.5 mmHg ( 20 °C)
InChI Key   ZTQSAGDEMFDKMZ-UHFFFAOYSA-N
assay   99%
autoignition temp.   390 °F
expl. lim.   12.5 %
refractive index   n20/D 1.380(lit.)
bp   75 °C(lit.)
mp   −96 °C(lit.)
density   0.8 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1129 - class 3 - PG 2 - Butyraldehyde
WGK Germany 
1
RTECS 
ES2275000
Flash Point(F) 
23 °F
Flash Point(C) 
-5 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?