EMAIL THIS PAGE TO A FRIEND
B103802 Aldrich

Butyronitrile

98%

Synonym: Propyl cyanide

Purchase

Properties

vapor density   2.4 (vs air)
vapor pressure   23 mmHg ( 25 °C)
InChI Key   KVNRLNFWIYMESJ-UHFFFAOYSA-N
assay   98%
autoignition temp.   910 °F
refractive index   n20/D 1.384(lit.)
bp   115-117 °C(lit.)
mp   −112 °C(lit.)
density   0.794 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2411 3/PG 2
WGK Germany 
1
RTECS 
ET8750000
Flash Point(F) 
64 °F
Flash Point(C) 
18 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

538264 Butyronitrile, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?