EMAIL THIS PAGE TO A FRIEND
B16305 Aldrich

Benzylamine

reagent grade

Synonym: α-Aminotoluene

Purchase

Properties

grade   reagent grade
InChI Key   WGQKYBSKWIADBV-UHFFFAOYSA-N
refractive index   n20/D 1.543(lit.)
bp   184-185 °C(lit.)
mp   10 °C(lit.)
density   0.981 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
1
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

185701 Benzylamine, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?