• USA Home
  • B28001 - 5-Benzyloxyindole-3-acetic acid

EMAIL THIS PAGE TO A FRIEND
B28001 Aldrich

5-Benzyloxyindole-3-acetic acid

98%

Synonym: NSC 68361

  • CAS Number 4382-53-0

  • Empirical Formula (Hill Notation) C17H15NO3

  • Molecular Weight 281.31

  •  MDL number MFCD00022750

Purchase

Properties

InChI Key   GKIOPUYLJUOZHJ-UHFFFAOYSA-N
assay   98%

Description

Application

Reactant for preparation of:
• Inhibitors of Gli1-mediated transcription as potential anticancer agents
• Synthetic analogs of spider toxins

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?