• USA Home
  • B4708 - Benzethonium chloride

EMAIL THIS PAGE TO A FRIEND
B4708 Aldrich

Benzethonium chloride

97%

Synonym: (Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride

  • CAS Number 121-54-0

  • Empirical Formula (Hill Notation) C27H42ClNO2

  • Molecular Weight 448.08

  •  Beilstein Registry Number 3898548

  •  EC Number 204-479-9

  •  MDL number MFCD00011742

  •  PubChem Substance ID 329773243

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   UREZNYTWGJKWBI-UHFFFAOYSA-M
assay   97%
mp   162-164 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2923 6.1(8) / PGIII
WGK Germany 
2
RTECS 
BO717500053752
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B8879 Benzethonium chloride, ≥97% (titration), ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?