EMAIL THIS PAGE TO A FRIEND
B54207 Aldrich

Biuret

97%

Synonym: Allophanic acid amide, Carbamoylurea

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks, Ureas More...
InChI Key   OHJMTUPIZMNBFR-UHFFFAOYSA-N
assay   97%
mp   185-190 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?