• USA Home
  • BR45550 - BRAND® evaporating dish
EMAIL THIS PAGE TO A FRIEND
BR45550 Aldrich
BRAND® evaporating dish
PTFE, volume 75 mL
Synonym: Evaporating dishes, PTFE
Purchase
Properties
Related Categories BRAND, BRAND Dishes, Labware, Manufacturer Browser
material   PTFE
  crucible
feature   without spout
mfr. no.   BRAND, 45550
volume   75 mL
Description
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?