• USA Home
  • C112151 - cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic dianhydride

EMAIL THIS PAGE TO A FRIEND
C112151 Aldrich

cis,cis,cis,cis-1,2,3,4-Cyclopentanetetracarboxylic dianhydride

96%

  • CAS Number 4802-47-5

  • Empirical Formula (Hill Notation) C9H6O6

  • Molecular Weight 210.14

Purchase

Properties

assay   96%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?