• USA Home
  • C55607 - 2-(4-Chloro-o-tolyloxy)propionic acid

EMAIL THIS PAGE TO A FRIEND
C55607 Aldrich

2-(4-Chloro-o-tolyloxy)propionic acid

98%

Synonym: α-(4-Chloro-2-methylphenoxy)propionic acid, CMPP

  • CAS Number 93-65-2

  • Empirical Formula (Hill Notation) C10H11ClO3

  • Molecular Weight 214.65

  •  EC Number 202-264-4

  •  PubChem Substance ID 329775094

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   WNTGYJSOUMFZEP-UHFFFAOYSA-N
assay   98%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN2811 - class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
UE9750000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?