• USA Home
  • C67206 - 3-Chlorophenyl methyl carbinol

EMAIL THIS PAGE TO A FRIEND
C67206 Aldrich

3-Chlorophenyl methyl carbinol

98%

Synonym: m-Chloro-α-methylbenzyl alcohol

Purchase

Properties

assay   98%

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?