• USA Home
  • CDS002431 - 4-Methyl-2-[(methylsulfonyl)amino]-1,3-thiazole-5-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
CDS002431 Aldrich

4-Methyl-2-[(methylsulfonyl)amino]-1,3-thiazole-5-carboxylic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C6H8N2O4S2

  • Molecular Weight 236.27

  •  MDL number MFCD06011074

  •  PubChem Substance ID 329779455

Purchase

Properties

Related Categories Building Blocks, C3 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More...
InChI Key   RDSIODJZQJVHRQ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?