• USA Home
  • CDS003379 - 2-methylsulfanylphenylboronic acid, pinacol ester

EMAIL THIS PAGE TO A FRIEND
CDS003379 Aldrich

2-methylsulfanylphenylboronic acid, pinacol ester

AldrichCPR

  • Empirical Formula (Hill Notation) C13H19BO2S

  • Molecular Weight 250.16

  •  MDL number MFCD05155219

  •  PubChem Substance ID 329780199

Purchase

Properties

Related Categories Aryl Boronate Esters, Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents More...
InChI Key   SLJWZEVOGXSTOM-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Price and Availability


KitAlysis - Increasing Catalysis Productivity
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Boronic Acid Esters - Chemfiles Volume 4 Article 2

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?