• USA Home
  • CDS004088 - (3-Methyl-2-oxo-2,3-dihydro-imidazo[1,2-a]pyridin-3-yl)-acetic acid

EMAIL THIS PAGE TO A FRIEND
CDS004088 Aldrich

(3-Methyl-2-oxo-2,3-dihydro-imidazo[1,2-a]pyridin-3-yl)-acetic acid

AldrichCPR

  • Empirical Formula (Hill Notation) C10H10N2O3

  • Molecular Weight 206.20

  •  MDL number MFCD03274736

  •  PubChem Substance ID 329780911

Purchase

Properties

Related Categories Amines, Building Blocks, C10, Chemical Synthesis, Nitrogen Compounds,
InChI Key   LDFNPELUPZWWBZ-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?