• USA Home
  • CDS021361 - rac-(1R,2S,3R,4S)-3-Amino-7-oxabicyclo[2.2.1]hept-5-ene-2-carboxylic acid HCl

EMAIL THIS PAGE TO A FRIEND
CDS021361 Aldrich

rac-(1R,2S,3R,4S)-3-Amino-7-oxabicyclo[2.2.1]hept-5-ene-2-carboxylic acid HCl

Synonym: rac-(1R,2S,3R,4S)-3-Amino-7-oxabicyclo[2.2.1]hept-5-ene-2-carboxylic acid hydrochloride

  • Empirical Formula (Hill Notation) C7H10ClNO3

  • Molecular Weight 191.61

  •  MDL number MFCD28043513

  •  PubChem Substance ID 329794625

Purchase

Properties

InChI Key   PLIWFZMNPKLWTL-HRDKBJBWSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?