• USA Home
  • CDS024585 - 1-[(tert-Butoxy)carbonyl]-2-ethylpyrrolidine-2-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
CDS024585 Aldrich

1-[(tert-Butoxy)carbonyl]-2-ethylpyrrolidine-2-carboxylic acid

  • Empirical Formula (Hill Notation) C12H21NO4

  • Molecular Weight 243.30

  •  MDL number MFCD23701808

  •  PubChem Substance ID 329796603

Purchase

Properties

InChI Key   KMOZPCFILJMJNU-UHFFFAOYSA-N

Description

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?