• USA Home
  • CLS606055 - Pyrex® Buchner funnel, with perforated plate
EMAIL THIS PAGE TO A FRIEND
CLS606055 Aldrich
Pyrex® Buchner funnel, with perforated plate
Purchase
Properties
Related Categories Buchner Funnels, Corning, Corning Buchner Filters, Corning Funnels, Funnels,
mfr. no.   Corning, 6060-55
diam.   55.0 mm
color   clear
Description
General description
With perforated plate. Suitable for vacuum filtration, but translucent plate should not be subjected to pressure differential in excess of 1atm.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?