• USA Home
  • CLS6950235 - Pyrex® vacuum manometers
EMAIL THIS PAGE TO A FRIEND
CLS6950235 Aldrich
Pyrex® vacuum manometers
Standard
Purchase
Description
General description
The manometer tube is enclosed in a round-bottom, graduated jacket and is sealed to a stopcock plug, that vents the outer jacket through a side tube in neck. Mercury is placed in the inner tube and in jacket bottom to measure the pressure differential. Not recommended for positive pressure. Red scale ranges from 0 to 160mm. With ST/NS: stopper. 235 × 25mm O.D. Mercury not included. Mercury-filling instructions are supplied with each manometer.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?