• USA Home
  • CLS80603 - Pyrex® centrifuge tubes with beaded rim, ungraduated
EMAIL THIS PAGE TO A FRIEND
CLS80603 Aldrich
Pyrex® centrifuge tubes with beaded rim, ungraduated
capacity 3.0 mL
Synonym: Corning® 8060
Purchase
Properties
Related Categories Corning, Corning Centrifuge Tubes, Corning Glass Centrifuge Tubes, Corning Tubes, Glass Tubes,
material   tube conical bottom
packaging   case of 12
  pack of 2
mfr. no.   Corning, 80603
O.D. × L   11 mm × 76 mm
capacity   3.0 mL
Description
General description
Formed from borosilicate glass. Stress that glass can withstand is greatly decreased by surface flaws such as scratches, chips, and cracks. Always use with correct sleeve or holder.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?