• USA Home
  • CLS88201924 - Pyrex® enlarging adapter
EMAIL THIS PAGE TO A FRIEND
CLS88201924 Aldrich
Pyrex® enlarging adapter
female Joint: ST/NS 19/38, male Joint: ST/NS 24/40
Purchase
Properties
Related Categories Corning, Corning Adapters, Enlarging/Reducing Adapters, Glassware, Glassware Adapters,
Joint   female Joint: ST/NS19/38
  male Joint: ST/NS24/40
mfr. no.   Corning, 8820-1924
availability   not available in EU
L   85 mm
Description
Legal Information
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?