• USA Home
  • CLS88202945 - Pyrex® enlarging adapter
EMAIL THIS PAGE TO A FRIEND
CLS88202945 Aldrich
Pyrex® enlarging adapter
female Joint: ST/NS 29/42, male Joint: ST/NS 45/50, L 85 mm
Purchase
Properties
Related Categories Corning, Corning Adapters, Enlarging/Reducing Adapters, Glassware, Glassware Adapters,
Joint   female Joint: ST/NS29/42
  male Joint: ST/NS45/50
mfr. no.   Corning, 8820-2945
availability   not available in EU
L   85 mm
Description
Legal Information
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?