• USA Home
  • CLS88252924 - Pyrex® bushing-type reducing adapter
EMAIL THIS PAGE TO A FRIEND
CLS88252924 Aldrich
Pyrex® bushing-type reducing adapter
female Joint: ST/NS 29/42, male Joint: ST/NS 24/40, L 56 mm
Synonym: glass adapter, glass adapters
Purchase
Properties
Related Categories Bushing-type, Corning, Corning Adapters, Enlarging/Reducing Adapters, Glassware,
Joint   female Joint: ST/NS29/42
  male Joint: ST/NS24/40
mfr. no.   Corning 8825
availability   not available in EU
L   56 mm
Description
General description
Heavy walls for mechanical strength and heavy upper rim for good finger grip. Shorter than conventional type for more compact assemblies. Useful for attaching jointed thermometers.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?