• USA Home
  • D135801 - (2,5-Dimethoxyphenyl)acetic acid

EMAIL THIS PAGE TO A FRIEND
D135801 Aldrich

(2,5-Dimethoxyphenyl)acetic acid

99%

Purchase

Properties

Related Categories Building Blocks, C10, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   BBZDYQUXRFATHZ-UHFFFAOYSA-N
assay   99%
mp   123-125 °C(lit.)

Description

Packaging

5 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?