EMAIL THIS PAGE TO A FRIEND
D178985 Aldrich

Dimethyl phthalate

99%

Synonym: DMP

  • CAS Number 131-11-3

  • Linear Formula C6H4-1,2-(CO2CH3)2

  • Molecular Weight 194.18

  •  Beilstein Registry Number 1911460

  •  EC Number 205-011-6

  •  MDL number MFCD00008425

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   NIQCNGHVCWTJSM-UHFFFAOYSA-N
assay   99%
autoignition temp.   1033 °F
refractive index   n20/D 1.515(lit.)
bp   282 °C(lit.)
mp   2 °C(lit.)
density   1.19 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
TI1575000
Flash Point(F) 
295 °F
Flash Point(C) 
146 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?