• USA Home
  • D219401 - 2,2′-Dithiodibenzoic acid

EMAIL THIS PAGE TO A FRIEND
D219401 Aldrich

2,2′-Dithiodibenzoic acid

97%

Synonym: 2-Carboxyphenyl disulfide, Bis(2-carboxyphenyl) disulfide, Dithiosalicylic acid

  • CAS Number 119-80-2

  • Linear Formula S2(C6H4CO2H)2

  • Molecular Weight 306.36

  •  Beilstein Registry Number 2221810

  •  MDL number MFCD00002465

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   LBEMXJWGHIEXRA-UHFFFAOYSA-N
assay   97%
mp   287-290 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?