• USA Home
  • D5057 - Dipropylenetriamine NONOate

EMAIL THIS PAGE TO A FRIEND
D5057 Aldrich

Dipropylenetriamine NONOate

Synonym: 3,3′-(Hydroxynitrosohydrazino)bis-1-propanamine, DPTA NONOate

Purchase

Description

Application

t1/2 NO release = 300 min (phosphate buffer at 22°C)

Biochem/physiol Actions

NO donor

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?