-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
Synonym: Propylene dichloride
CAS Number 78-87-5
Linear Formula CH3CHClCH2Cl
Molecular Weight 112.99
Beilstein Registry Number 1718880
EC Number 201-152-2
MDL number MFCD00000868
PubChem Substance ID 24894090
| Related Categories | Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More... |
| vapor density | 3.89 (vs air) |
| vapor pressure | 40 mmHg ( 19.4 °C) |
| assay | 99% |
| autoignition temp. | 1035 °F |
| expl. lim. | 14.5 % |
| refractive index |
n |
| bp | 95-96 °C(lit.) |
25, 100 g in glass btl
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?