EMAIL THIS PAGE TO A FRIEND
E43209 Aldrich

Ethyl oxamate

98%

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   RZMZBHSKPLVQCP-UHFFFAOYSA-N
assay   98%
mp   113-116 °C(lit.)

Description

Packaging

100 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?