EMAIL THIS PAGE TO A FRIEND
E44108 Aldrich

3-Ethylphenol

99%

  • CAS Number 620-17-7

  • Linear Formula C2H5C6H4OH

  • Molecular Weight 122.16

  •  Beilstein Registry Number 1857002

  •  MDL number MFCD00002311

Purchase

Properties

InChI Key   HMNKTRSOROOSPP-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.533(lit.)
density   1.001 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?