• USA Home
  • G5000 - DL-Glyceric acid hemicalcium salt hydrate

EMAIL THIS PAGE TO A FRIEND
G5000 Aldrich

DL-Glyceric acid hemicalcium salt hydrate

99%

Synonym: 2,3-Dihydroxypropanoic acid hemicalcium salt, Hemicalcium DL-glycerate, hemi-Calcium DL-glycerate

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   OKEVQKKVYQHNFW-UHFFFAOYSA-L
assay   99%
mp   134 °C (dec.)(lit.)

Description

Packaging

1 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

G7274 DL-Glyceric acid hemicalcium salt hydrate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?