EMAIL THIS PAGE TO A FRIEND
G7801 Aldrich

Gly-Gly

98%

Synonym: Diglycine, Glycyl-glycine

  • CAS Number 556-50-3

  • Linear Formula NH2CH2CONHCH2COOH

  • Molecular Weight 132.12

  •  Beilstein Registry Number 1765223

  •  EC Number 209-127-8

  •  MDL number MFCD00008130

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   YMAWOPBAYDPSLA-UHFFFAOYSA-N
assay   98%
useful pH range   7.5 - 8.9
pKa    8.2

Description

Application

Buffer useful in the physiological pH range. pKa=8.40 at 20°C. Used in peptide modeling and drug solubilizing studies.

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?