• USA Home
  • GF02694618 - Polyamide - Nylon 6, 6

EMAIL THIS PAGE TO A FRIEND
GF02694618 Aldrich

Polyamide - Nylon 6, 6

(PA 6,6), sheet, thickness 5 mm, size 300 × 300 mm

Synonym: AM323050

Purchase

Properties

form   sheet
mfr. no.   Goodfellow, 026-946-18
size   300 × 300 mm
thickness   5 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?