EMAIL THIS PAGE TO A FRIEND
GF05942987 Aldrich

Polyimide

(PI), laminate, thickness 0.06 mm, L 1 m, polymer thickness 0.025mm

Synonym: IM301522

Purchase

Properties

mfr. no.   Goodfellow, 059-429-87
L   1 m
thickness   0.06 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?