• USA Home
  • GF14008867 - Polymethylmethacrylate

EMAIL THIS PAGE TO A FRIEND
GF14008867 Aldrich

Polymethylmethacrylate

(PMMA, ACRYLIC), sheet, thickness 3 mm, size 150 × 150 mm, condition cast

Synonym: ME303032

Purchase

Properties

form   sheet
mfr. no.   Goodfellow, 140-088-67
size   150 × 150 mm
thickness   3 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?