• USA Home
  • GF15475811 - Polyethersulfone

EMAIL THIS PAGE TO A FRIEND
GF15475811 Aldrich

Polyethersulfone

(PES), rod, diam. 12.7 mm, L 100 mm

Synonym: SU307913

Purchase

Properties

form   rod
mfr. no.   Goodfellow, 154-758-11
L   100 mm
diam.   12.7 mm

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?