• USA Home
  • GF18928525 - Polyethersulfone

EMAIL THIS PAGE TO A FRIEND
GF18928525 Aldrich

Polyethersulfone

(PES), sheet, thickness 2 mm, size 102 × 102 mm, clear amber

Synonym: SU303200

Purchase

Properties

form   sheet
mfr. no.   Goodfellow, 189-285-25
size   102 × 102 mm
thickness   2 mm
color   clear amber

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?