• USA Home
  • GF27595446 - Polyethylene - Medium Density

EMAIL THIS PAGE TO A FRIEND
GF27595446 Aldrich

Polyethylene - Medium Density

(MDPE), powder, max. particle size 350micron, weight 200 g

Synonym: EV306010

Purchase

Properties

form   powder
mfr. no.   Goodfellow, 275-954-46
weight   200 g

Description

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?